Chemware® Dipper, Teflon® PFA, Saint-Gobain Performance Plastics
# de catalogue 56925-300
Fournisseur: Saint Gobain Performance Plastics
Spécifications
- Description:Chemware® Dipper
- Cat. no.:56925-300
- Supplier No.:D1069102
Spécifications
A propos de cet article
Use for ultra-pure applications and with aggressive media
Handle hooks over edge of container, eliminating dripping of corrosive fluids onto surfaces. The 6.4 cm (21/2") diameter allows for use with containers having at least a 7.6 cm (3") opening and a capacity ranging from 300 to 1000ml.
Customers Who Bought This Also Bought