Glass Coliwasa Samplers, Wheaton®, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Ideal for use in the collection of environmental and industrial samples
These borosilicate glass samplers are specifically designed for hazardous waste and groundwater applications and can be used to obtain samples from drums, tank cars, lagoons, lakes, rivers, and storage tanks. Samplers are available unscored for repeated use, or prescored for disposal after a single use. They feature liquid-tight PTFE seals that lock the sample column into place and a sample capacity of 200mL. Samplers measure 2.2dia.x106.7Lcm (7/8x42") and have a tapered bottom with a 1.3cm (1/2") diameter opening.
Ordering information: Replacement seals for unscored or prescored samplers are sold separately.
Customers Who Bought This Also Bought