Sample Thieves (Bacon Bombs), Koehler
Fournisseur: Koehler
Certificats
A propos de cet article
Devices obtain samples from any level in storage tanks, tank cars, and drums
A plunger assembly opens to admit the sample when the thief strikes the bottom of the tank or is triggered by a secondary trip line. Plunger locking cam provides a tight closure during transport, except for 2.8cm (11/8") diameter models, which are designed for sampling through narrow pipes and openings.
Customers Who Bought This Also Bought