Coliwasa Sampler, Glass, Disposable, Wheaton®, DWK Life Sciences
# de catalogue 66174-220
Fournisseur: DWK LIFE SCIENCES S. DE R.L. DE MX
Small Business Enterprise
Spécifications
- Description:Coliwasa Sampler
- Cat. no.:66174-220
- Supplier No.:885220
Spécifications
A propos de cet article
For liquid waste sampling
Takes a 200mL sample from a full drum. Inner and outer tubes are made from soda lime glass and are prescored for easy disposal. Unit measures 106.7Lx2.2dia.cm (42x7/8"). Tapered bottom has 1.3cm (1/2") diameter opening.
Customers Who Bought This Also Bought