ContactUsLinkComponent
Bomb Sampler, Stainless Steel, Wheaton®, DWK Life Sciences
Bomb Sampler, Stainless Steel, Wheaton®, DWK Life Sciences
# de catalogue 66174-200
Small Business Enterprise
Bomb Sampler
Bomb Sampler, Stainless Steel, Wheaton®, DWK Life Sciences
# de catalogue 66174-200
Small Business Enterprise
Spécifications
  • Description:
    Bomb sampler, type 304 stainless steel
  • Cat. no.:
    66174-200
  • Supplier No.:
    885200
Spécifications
A propos de cet article
Efficient composite sampling device for collecting liquid samples from storage tanks, tank cars, ponds, lagoons and wells
Type 304 stainless steel. Size: 6.35 dia.×25.4L cm (2¹/₂×10"). Capacity: 500 ml (17 oz.). Weight: 1.8 kg (4 lbs.). Helps to comply with EPA Composite Sampling Method 1.1.4.1.
May be activated at various depths. Consists of cylindrical reservoir chamber, a weighted plunger that seals the chamber at bottom, and cable attachments for suspending the apparatus and activating the sampling device. (Cable not included.)
Customers Who Bought This Also Bought