Coliwasa Samplers, PVC, Reusable, Wheaton®, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
For safe and simple sampling of drums and tanks.
Uniquely designed top spin bar and bottom valve plate mechanism produce a positive seal for sample retention. Provides excellent control for releasing the sample into containers. Modular design adds flexibility by allowing the connection of several samplers to form a sampling unit up to 20 feet in length via connectors (sold separately). Samplers are chemically-resistant, gray colored PVC.
Ordering information: Samplers and connectors (sold separately) available in 121.9 cm (48") and 182.9 cm (72") lengths. Samplers and connectors each supplied with a 121.9 cm (48") long cleaning brush.
Customers Who Bought This Also Bought