XRF Thin-Film Sample Supports, Chemplex®
Fournisseur: Chemplex
Certificats
A propos de cet article
For retaining liquid, powdered, slurry or solid specimens in XRF Sample Cups for wavelength (WDXRF) and energy dispersive (EDXRF) spectrochemical analysis.
- Chemplex thin-film sample support substances are manufactured to very close tolerances (1 to 2% within a single package)
Choose from Prolene, ultra-polyester, polypropylene, Mylar, polycarbonate or polyimide support materials based on their ability to safely retain specimen during preparation and analysis stages, permit maximum analyte-line transmittance, and provide statistically reproducible analytical data.
Considerations for selecting an appropriate thin-film material include: resistance of material to degradation by the analyte, analyte concentration/wavelength, thin-film purity and thin-film thickness uniformity.
Available in continuous rolls, precut squares and circles.
Customers Who Bought This Also Bought