Liquid-Binder™ Aqueous Additive, Chemplex®
# de catalogue 15566-086
Fournisseur: Chemplex
Numéro CAS:
Spécifications
- Description:Liquid-Binder™ Aqueous Additive
- Size:480 cc (1 pt.)
- Cat. no.:15566-086
- Supplier No.:800
Spécifications
A propos de cet article
Converts powdered samples into a slurry to eliminate segregation by size or density during comminution process.
Particularly useful for compositionally complex powders consisting of particles of different size, shape, density and hardness. Ideal for XRD analysis where Liquid-Binder™ is useful in forming self-supporting cylindrical samples for direct mounting in a camera. May also be used as a dispersing/mounting medium.
Customers Who Bought This Also Bought