Boric Acid Tablets, Chemplex
# de catalogue 15566-084
Fournisseur: Chemplex
Numéro CAS:
Spécifications
- Packaging:1000 Tablets/Bottle
- Description:Boric Acid Tablets
- Size:0.5 g Tablets
- Cat. no.:15566-084
- Supplier No.:750
Spécifications
A propos de cet article
Very abrasive heat-resistant additive suitable for processing samples that tend to reweld during comminution in metallic grinding vessels.
Frequently used in conjunction with x-ray mix to impart lubricity for “plasticizing” pellets.
Customers Who Bought This Also Bought