SpectroVial® Sample Grinding And Blending Vials, Chemplex Industries
Fournisseur: Chemplex
Certificats
A propos de cet article
Comminuting and blending vials specially designed for spectrochemical powdered sample material processing in ball pestle impact grinders.
Concavely contoured interior ends in both main cylindrical body and closure promote particle-size reduction.
Delivery: Titanium carbide SpectroVials® are supplied with tungsten carbide ball pestles; agate SpectroVials® with agate ball pestles; stainless steel SpectroVials® with stainless steel ball pestles; and polystyrene SpectroVials® with methyl metacrylate ball pestles.
Customers Who Bought This Also Bought