A propos de cet article
Photoluminescent exit signs are steel-encased glow-in-the-dark plastic panels
Three types of framed exit signs are available: single-face wall-mounted, single-face with mounting bracket, or double-face with mounting bracket. Signs come in three colors: black, red, and green. Extra brackets are also available.
These signs provide consistent and highly visible illumination immediately after the lights go out. Signs absorb and store energy from ambient light. They use zero electricity and do not require special wiring, maintenance, a backup power source, or professional installation. Punch-out or field-applied chevrons provide direction. Signs are easily installed and are perfect for high and low level applications. Intended for indoor use only. Compliant with all current building codes.