Spécifications
- Capacity:2.5 L
- Color:Amber
- Description:24-Hour Urine Collection Container
- Cat. no.:14005-980
- Packaging:40/Pack; 40/Case
- Supplier No.:30 2500
Spécifications
A propos de cet article
Alleviate mess and exposure when handling urine collection and sampling with the Thermo Scientific Samco 24-hour Urine Collection product line
Alleviate mess and exposure when handling urine collection and sampling with the Thermo Scientific Samco 24-hour Urine Collection product line
These HDPE containers are amber tinted to protect from light exposure and are inert to normal preservatives.