96-Well Microplate Orienter Stickers, Diversified Biotech
Catalogue # 89032-141
Fournisseur: Diversified Biotech
Small Business Enterprise
Spécifications
- Description:96-well microplate orienter stickers
- Cat. no.:89032-141
- Supplier No.:WEST-1000
Spécifications
À propos de cet article
Use stickers to permanently label multiple microplates.
- Made of clear polyester with acrylic adhesive
Each well is clearly marked with a corresponding row number and column letter for easy cross-referencing. The paper backing is scored in the middle for easy aligning on the underside of the plate.