Warning Label 'Formaldehyde', Shamrock
# de catalogue 36436-648
Fournisseur: Shamrock Scientific
Small Business Enterprise
Spécifications
- Description:Warning labels
- Cat. no.:36436-648
- Legend:Formaldehyde
- Dimensions:44.5×63.5 mm (1.75×2.5")
- Supplier No.:SB-386H
Spécifications
A propos de cet article
Designed to identify containers or vessels that contain formaldehyde.
- Strong adhesive sticks tightly to glass, plastic, metal, and wood
A white background with bold black lettering allows for quick, simple identification. 'Caution Contains Formaldehyde' is printed above a description of exposure symptoms.
Customers Who Bought This Also Bought