Specimen and Pathology Tape Labels, TimeMed
Fournisseur: Time Med Labeling Systems
Certificats
A propos de cet article
Designed to assist in patient identification and safety.
- Minimize the possibility of transmitting diseases
- Ensure proper specimen identification
Label 36437-368 has space for Name, Hosp. No., Rm. No., and Date.
Label 36437-374 has space for Name, Date, Sex, Age, Room, Doctor, Specimen, and Number.
White labels printed in black.
Ordering information: All labels are supplied on an individual roll containing 12.7 m (500") of tape. Label 36437-368 feature 222 imprints per roll. Label 36437-374 features 143 imprints per roll.
Customers Who Bought This Also Bought