Write-On Hazardous Communication Labels, National Marker
Fournisseur: ACCUFORM MANUFACTURING, INC
Certificats
A propos de cet article
Customize warnings in the workplace with these pressure-sensitive write-on labels.
Available in paper and two configurations of vinyl labels. (vinyl label provides space for protective equipment symbol usage.) The 0.004 mm thick vinyl labels are printed in fade-resistant UV ink.
Ordering information: Use the vinyl penetrating pen on vinyl labels, and the erasable dry maker pen on paper labels (both sold separately).
Customers Who Bought This Also Bought