Shipping Labels, DOT, National Marker
Fournisseur: ACCUFORM MANUFACTURING, INC
Certificats
A propos de cet article
Pressure-sensitive paper labels comply with DOT HM-181 specifications when shipping hazardous materials (DOT 49 CFR 172.407).
Ordering information: Also available as 0.004 mm pressure-sensitive vinyl labels; contact your Avantor representative for details.
Customers Who Bought This Also Bought