I-CHEM™ Color-Coded Environmental Sample Labels, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Designed for efficient sample handling and identification in the field and laboratory.
Self-adhesive paper labels have a 25.4mm (1") diameter; 1000 labels/roll.
Self-adhesive paper labels have a 25.4 mm (1") diameter; 1000 labels/roll.
Labels are color-coded with standard preservative types for quick reference. Use the plain label to create a customized label.
Customers Who Bought This Also Bought