Sani-S'wipe Sterile Sampling Kits
Fournisseur: Labplas
New Product
Certificats
A propos de cet article
The Sani-S'wipe is a kit comprising a blue cloth and a sterile bag.
- ISO 18593 compliant
- Time-saving solution
- Ideal for various surfaces
- Certified sterile
- Integrated closure system
- Biocide-free cloth for sampling
- Thermoplastic laminate material
- Transparent
The Sani-S'wipe is a kit comprising a blue cloth and a sterile bag, designed for the collection of samples used to detect the presence of microbiological contaminants and other foodborne pathogens on various surfaces. It is easy to use and is ideal for large areas. It guarantees an accurate representation of the controlled surface in one quick movement.
Dimensions: 190×292 mm
Thickness: 3.5 mil
Customers Who Bought This Also Bought