Labels 'Hazardous Materials', National Marker
# de catalogue 36434-364
Fournisseur: ACCUFORM MANUFACTURING, INC
Spécifications
- Description:Labels for hazardous materials
- Cat. no.:36434-364
- Legend:Caution hazardous material storage area
- Dimensions:76.2×127 mm (3×5")
- Supplier No.:C310AP
Spécifications
A propos de cet article
Use these labels to draw attention to hazards associated with equipment, machinery, and storage areas.
Pressure-sensitive vinyl adheres to almost any surface, rough or smooth. Labels can be written on to provide additional information as desired. For indoor or outdoor use.
Customers Who Bought This Also Bought