Davis Instruments Vantage Pro2 Wireless Weather Station
# de catalogue 470356-492
Fournisseur: Davis Instruments
New Product
Spécifications
- Description:Vantage Pro2
Spécifications
A propos de cet article
The davis vantage pro2, a research-grade weather station, is designed to deliver reliable, accurate data year after year and is highly configurable, coming with a wide range of sensors and the ability to add on others. It also provides historical data, highs and lows, moon phase, indexes, forecasts and much more.
- Accurate, reliable weather monitoring with real-time data updates every 2.5 seconds
- Industry-leading transmission range of 1,000’/300 m from the sensor suite to the console
- Sensor suite’s rugged, weather-proof housing gives you years of reliable data
- Sensor suite includes outside temperature and humidity sensors in a passive radiation shield; wind speed and direction; and rainfall
- Mount anemometer up to 40’/12 m from the rest of sensor suite for flexible siting
- Includes WeatherLink Console with the vibrant, HD color touchscreen
- Sensor suite is solar-powered with battery back-up; console is AC-powered with battery backup
It reports real-time wind speeds up to 200 mph, temperature and humidity, barometric pressure, rainfall and other information such as heat index, dew point, highs and lows, and much more. Users have the option to include solar and UV radiation sensors, as well as a fan-aspirated radiation shield. Vantage Pro2 sensors are NIST-traceable and are suitable for professional weather data reporting.
Customers Who Bought This Also Bought