Shoe Brush Machine, 2001TB, Liberty Industries
# de catalogue 21924-463
Fournisseur: Liberty Industries
Small Business Enterprise
Spécifications
- Description:Shoe Brush Machine
- Cat. no.:21924-463
- Supplier No.:20-2240
Spécifications
A propos de cet article
This shoe brush machine is designed with four transmission-driven brushes, including a floating weight-loaded top brush and rotating bottom brush that clean sides, top, and bottom of shoes prior to entry into contamination-controlled areas.
Rubber shin guard and cast iron base
Safety control switch and instructions are located on the support handle to assist in maintaining balance
white polyethylene cover with a removable hood secured by four latches
For 110 V, 60 Hz. 1/3 hp motor
Dimension: 51 x 47 x 99 cm (20 x 18 1/2x 39")
Certifications: Complies with U.S. Air Force Tech. Order T.O. 00-25-203 and Federal Standard 209.
Ordering information: Machine should be used with a vacuum system, sold separately. Unit dimensions are given with support handle included.
Customers Who Bought This Also Bought