VWR® Slide Labels
Fournisseur: VWR International
Certificats
A propos de cet article
Designed to help prevent curling or detaching, slide labels are printed on smudgeproof litho sheets and feature a permanent adhesive.
- Labels have a thickness 0.012 cm (0.0078")
- Color: white
Designed to help prevent curling or detaching, slide labels are printed on smudgeproof litho sheets and feature a permanent adhesive.
Customers Who Bought This Also Bought