VWR® Write-On/Wipe-Off Magnetic Shelf Label Rolls
Fournisseur: VWR International
Certificats
A propos de cet article
Magnetic label material has a white vinyl surface that may be written on with wet-erase marker and then wiped off.
- May be cut down to any size
Provided in a rol. Thickness: 0.8 mm (¹/₃₂"). Roll length: 15.2 m (50').
Customers Who Bought This Also Bought