ContactUsLinkComponent
Nalgene® PolyPaper® Right-To-Know Custom Labeling System, Thermo Scientific
Nalgene® PolyPaper® Right-To-Know Custom Labeling System, Thermo Scientific
# de catalogue 36471-606
PolyPaper* Right-to-Know Custom Labeling System, NALGENE*
Nalgene® PolyPaper® Right-To-Know Custom Labeling System, Thermo Scientific
# de catalogue 36471-606
Spécifications
  • Description:
    Right-to-know custom labeling system
  • Material:
    Ploypaper
  • Cat. no.:
    36471-606
  • Dimensions:
    76.2×127 mm (3×5")
  • Supplier No.:
    6316-1000
Spécifications
A propos de cet article
Pressure-sensitive labels incorporate Department of Transportation and National Fire Protection Association codes, symbols, and pictorials of recommended safety equipment in an easily recognizable format.
Custom Labeling System includes pressure sensitive stickers depicting NFPA rating numbers, hazard codes, and safety pictorials to be placed on the blank PolyPaper Label. Assures compliance with the OSHA Hazard Communication Regulation CFR 29 1910.1200 (f)(4).
  • Made of tear-resistant NALGENE® PolyPaper
  • Waterproof and chemical resistant
Pressure-sensitive labels incorporate Department of Transportation and National Fire Protection Association codes, symbols, and pictorials of recommended safety equipment in an easily recognizable format.
Custom Labeling System includes pressure sensitive stickers depicting NFPA rating numbers, hazard codes, and safety pictorials to be placed on the blank PolyPaper Label.
A Mylar® overlay is provided to protect the label from chemicals. A special adhesive backing adheres to any plastic, glass or metal transfer container.
Certifications: Assures compliance with the OSHA Hazard Communication Regulation CFR 29 1910.1200 (f)(4).
Customers Who Bought This Also Bought