Satin Cloth Labels, Shamrock
Fournisseur: PCS/AI,INC MX
Certificats
A propos de cet article
Ideal for cryogenic procedures that use dry ice, liquid nitrogen, or deep freezing.
Labels withstand extremely cold temperatures. Rayon cloth resists acids, alkalies, oils, grease, soap, and fresh or salt water. Textured surface is easily marked with pen, pencil, or felt-tip marker. Pressure sensitive backing adheres to almost any surface.
Customers Who Bought This Also Bought