ContactUsLinkComponent
VWR® Write-On Metal Tags
VWR® Write-On Metal Tags
# de catalogue 36419-213
Fournisseur:  VWR International
VWR* Write-on Metal Tags
VWR® Write-On Metal Tags
# de catalogue 36419-213
Fournisseur:  VWR International
Spécifications
  • Description:
    Write-on metal tags
  • Material:
    Metal
  • Cat. no.:
    36419-213
  • Dimensions:
    34.9×79.5 mm (1.38×3.13")
Spécifications
A propos de cet article
Permanently engrave your message on this metal tag with an ordinary ballpoint pen.
  • Extremely durable
  • Suited to extreme hot or cold temperatures
Permanently engrave your message on this metal tag with an ordinary ballpoint pen.
The aluminum tag features a 'soft center' and will take messages on both sides.
Delivery: Package of 48 tags with wire fasteners.
Customers Who Bought This Also Bought