SP Wilmad-LabGlass Flash Chromatography Reservoirs, SP Industries
Supplier: Avantor
Certificates
About this item
These reservoirs are used when larger volumes are required to elute samples from flash chromatography column.
- Borosilicate glass
- For containment of mobile phase
- Connects flow controller and column
Upper outer joint connects to flow controller while lower inner joint connects with column.
Customers Who Bought This Also Bought