Blot Paper, NuSep®
# de catalogue 16003-896
Fournisseur: NuSep
Spécifications
- Dimensions:7 x 8.5 cm (23/4 x 311/32")
- Description:For Wet Blots
- Cat. no.:16003-896
- Supplier No.:RB-300
Spécifications
A propos de cet article
Thin paper for wet blots and thick paper for semi-dry blots is 7x8.5cm (23/4x35/16").
Customers Who Bought This Also Bought