KONTES® ULTRA-WARE® Safety System 3, Kimble Chase
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
This cap system eliminates the safety hazards associated with aluminum foil-wrapped solvent containers
The system includes a three-hole PTFE cap, PEEK™ combination inlet filter and sparger, ETFE flangeless nuts and ferrules, ETFE vent port plug, and FEP fluoropolymer resin tubing. Systems are available with or without a solvent bottle adapter that converts GL-45 mobile phase caps to standard 4L bottles for direct connection to an HPLC pump.
Customers Who Bought This Also Bought