Crews® Blackjack® Protective Eyewear, MCR Safety
Supplier: MCR Safety
Certificates
About this item
These glasses feature a single, dual-focus wraparound lens design.
- Integral side shields for added protection
- Metal temples with spring hinge design provide maximum flexibility
Strong, lightweight, polycarbonate lenses with Duramass® scratch-resistant coating filter out 99% of UV radiation.
Certifications: Meets or exceeds ANSI Z87.1+ standards
Customers Who Bought This Also Bought