1400 Series Visitor Protective Eyewear, Encon® Safety Products
# de catalogue 46500-071
Fournisseur: Encon Safety Products
CSA Certified
Spécifications
- Lens Tint:Clear
- Cat. no.:46500-071
- Supplier No.:05148002
Spécifications
A propos de cet article
Eyewear is flexible enough for comfort during extended use and strong enough to provide maximum impact protection.
- One-piece spherical lens design offers 99% ultraviolet filtration
- Equipped with polycarbonate side shields for high-impact absorption
Glasses feature a double pin hinge for added durability, and a universal nose bridge for a snug, secure fit.
Certifications: Meet ANSI Z87.1-2003 and CSA Z94.3-2002 protection standards. CE marked.
Customers Who Bought This Also Bought