Rheodyne® Stainless Steel Sample Loops, IDEX Health & Science
Fournisseur: Rheodyne
Certificats
A propos de cet article
For Rheodyne® injector models 7125 and 7010.
Stainless steel loops are supplied with unswaged fittings so the tube can be completely bottomed in the injection port before the ferrule is swaged. The loop volumes are nominal. The actual volumes can differ from the nominal designations because of the ± 0.001" tolerance of the tubing bore. Accuracy of large loops (0.040" bore) is about 5%, intermediate loops (0.020") 10%, and small loops (0.007") 30%.
Customers Who Bought This Also Bought