A propos de cet article
Upchurch Scientific® Back Pressure Regulators (BPRs) are designed to enhance system performance through outgassing prevention and improved pump check valve efficiency.
Choose from a line of Biocompatible (PEEK™ body) and Stainless Steel BPR Assemblies, each complete with a replaceable, factory preset cartridge (except 5psi version). Upchurch Scientific® Biocompatible and Stainless Steel BPR Assemblies create incremental back pressures ranging from 5 to 1,000psi (0.3 to 69 bar).
Replacement BPR cartridges will operate in any of the standard BPR holders. The recommended operating flow rate is 0.1 to 10 mL/min. Within this range, the amount of back pressure created by the BPR Cartridge and Assemblies will not vary more than ±10%. Lower or higher flow rates may result in larger pressure fluctuations.
The following materials are used in these products; BPR cartridges are PEEK™, ETFE, PCTFE, Perfluoroelastomer, and Gold-Plated Stainless Steel. Biocompatible BPR Assemblies include these same materials (53521-124 also includes unplated stainless steel). Stainless Steel Assemblies include all of the BPR cartridge materials plus Delrin®.
Spécifications
- Catalog No:
- 21001-206
- 21511-598
- 97055-106
- 53521-124
- 21511-600
- 21511-588
- 21511-590
- 53521-132
- 21511-596
- 21511-586
- 21528-210
- BPR assembly back pressure rating:
- 2000 psi (138 bar)
- 2000 psi (138 bar)
- 2500 psi (172 bar)
- 2000 psi (138 bar)
- 2000 psi (138 bar)
- 2000 psi (138 bar)