Replacement Parts for Injectors and Switching Valves, Rheodyne®
Fournisseur: Rheodyne
Certificats
A propos de cet article
Vespel® rotor seals have an operating pH range from 0 to 10
Tefzel® rotor seals have a pH range from 0 to 14. Vespel® seals are recommended, except when operating at high pH.
Customers Who Bought This Also Bought