Static Free™ Hand Guard® Lotion, ITW Chemtronics
# de catalogue 15145-450
Fournisseur: ITW Chemtronics
Spécifications
- Capacity:237 ml (8 oz.)
- Description:Pump Bottle
- Supplier No.:C805
Spécifications
A propos de cet article
This hand lotion is formulated to protect against electrostatic charge
Nongreasy formula contains aloe vera and Vitamin E. Contains no lanolin, silicone, or mineral oil that could inhibit solderability of printed circuits or components. The emollients penetrate deeply to moisten and refresh dry, chapped, or chemically irritated skin.
Customers Who Bought This Also Bought