RheFlex® PEEK™ Fittings, Rheodyne®, IDEX Health & Science
Fournisseur: Rheodyne
Certificats
A propos de cet article
RheFlex® fittings provide inert nonmetal connections to 10-32 internal thread ports, including all Rheodyne® injectors, valves, and sample loops
Nuts and ferrules do not bind to each other during tightening and are reusable. The standard fitting can be wrench or finger tightened, and the short fitting is wrench tightened only.
The 1/16" fitting will hold PEEK, stainless steel, and titanium tubing to 5000psi.
Ordering information: Set contains five nuts and five ferrules.
Customers Who Bought This Also Bought