Spécifications
- Color:White
- Description:Preemie heel stick devices
- Incision Depth:0.085 cm
- Incision Length:0.175 cm
- Supplier No.:TFP50I
Spécifications
A propos de cet article
These single-use heel-stick devices feature a highly sharpened surgical blade that swings across the skin in a gentle arc.
- Sterile
- Choice of devices suitable for preemies (low birth weight) or newborns (birth - six months)
The incision produced is through the rich capillary bed, but is carefully controlled to a depth that is superficial to most nerve fibers. The result is a high quality blood sample with significantly less pain and trauma to the skin.
Once the blade is triggered, it retracts permanently back into the device, thereby reducing the possibility of accidental sticks. The need for repeat sticks is virtually eliminated because each device is carefully controlled to produce an incision at a standardized length and depth.