Spill Containment Trays, Scientific Plastics
Fournisseur: Scientific Plastics
Certificats
A propos de cet article
One-piece, rotationally-molded polyolefin trays protect cabinets and shelving from acid spills
Corners are rounded for easy cleaning. Walls are 4.8mm (3/16") thick for rigidity. Dishwasher safe. Depth: 2.4cm (1"). Color: natural.
Customers Who Bought This Also Bought