Synthware Adapter, Claisen, Threaded, Microscale, Kemtech America
# de catalogue 60004-112
Fournisseur: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- ST Joint:14/10
- Supplier No.:A371410
Spécifications
A propos de cet article
Claisen joint adapter for microscale applications
- Threaded
- For microscale applications
- Two compression caps and two O-rings included
Threaded Claisen joint adapter is supplied with two nylon compression caps and two Viton® O-rings.
Supplied with two nylon compression caps and two Viton® O-rings.
Customers Who Bought This Also Bought