Synthware Adapter, Distillation, Threaded, Microscale, Kemtech America
Catalog # 60004-110
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- ST Joint:14/10
- Cat. no.:60004-110
- Supplier No.:A351410
Specifications
About this item
Distillation joint adapter is suitable for microscale applications.
- Threaded
- For microscale applications
- Compression cap and O-ring included
Threaded distillation joint adapter is supplied with a nylon compression cap and Viton® O-ring.
Customers Who Bought This Also Bought