Synthware Valve, High Vacuum, Metering, PTFE-Protected, Low Hold Up Sidearms
Fournisseur: Kemtech America
Certificats
A propos de cet article
High vacuum metering valve with two 9mm O.D
- Hi vacuum valves
- Heavy wall borosilicate construction
- Teflon® protected
sidearms bent to minimize hold up of liquids. The Viton® O-rings on the PTFE plug shaft are protected with precision PTFE wipers. The tip of the plug has an integral O-ring to make the primary seal. This design provides an excellent vacuum-tight seal with PTFE as the only material exposed to chemicals.
Customers Who Bought This Also Bought