Synthware Valve, High Vacuum, PTFE-Protected, 180° Sidearms, Kemtech America
Fournisseur: Kemtech America
Certificats
A propos de cet article
High vacuum valve with two 9 mm O.D.
- Hi Vacuum valves
- Heavy wall Borosilicate construction
- Teflon® Protected
sidearms sealed to the barrel at a 180° angle. The Viton® O-rings on the PTFE plug shaft are protected with precision PTFE wipers. The tip of the plug has an integral O-ring to make the primary seal. This design provides an excellent vacuum-tight seal with PTFE as the only material exposed to chemicals.
Customers Who Bought This Also Bought