Wall-Mounted Gowning Racks, Advance Tabco®
Fournisseur: Advance Tabco
Certificats
A propos de cet article
Use in cleanroom gowning areas for storing garments.
- Hanger bar is scored in 5.1 cm (2") increments and protrudes 45.7 cm (18") from wall
- Overshelf allows for extra storage
Constructed from 16-gauge, type 304 stainless steel with a tubular design for easy assembly and laminar flow.
Ordering information: Each rack is supplied with twelve closed-loop stainless steel electropolished hangers. Shipped knocked down.
Customers Who Bought This Also Bought