Stainless Steel Gowning Benches, Advance Tabco®
Supplier: Advance Tabco
Certificates
About this item
Durable, corrosion-resistant gowning benches provide sturdy support for workers dressing for the cleanroom environment.
- Rolled rim front and back for worker comfort during changing
Constructed of 16-gauge, Type 304 stainless steel. Seat is 30.5 cm (12") wide; bench is 45.7 cm (18") high. Bench’s unique 'rainbow' leg design and 2.5 cm (1") adjustable plastic bullet feet permit freestanding use.
Ordering information: Benches can also be adapted for more secure floor mounting by ordering optional stainless steel flange feet. Units can be electropolished for an additional charge. Contact your Avantor sales representative for more information.
Customers Who Bought This Also Bought