Freestanding Gowning Racks, Advance Tabco®
Supplier: Advance Tabco
Certificates
About this item
Freestanding garment rack is made from 16-gauge, Type 304 stainless steel.
- Cross-bracing enhances rigidity
- Ideal for cleanroom gowning areas
Tubular design for easy assembly and laminar flow. Hanger bar is scored in 5.1 cm (2") increments. All units are 61 cm (24") wide. Cross-bracing enhances rigidity. Stainless steel flange feet for permanent floor mounting are available separately.
Ordering information: Supplied with twelve closed-loop stainless steel electropolished hangers. Supplied with 2.5 cm (1") adjustable plastic bullet feet. Shipped knocked down.
Frequently Bought Together