Spécifications
- Description:Lens Cleaning Station
- Cat. no.:52848-152
- Supplier No.:248350000
Spécifications
A propos de cet article
Designed for economy and convenience, this self-contained, disposable station combines cleaning solution and lens tissue.
- Disposable station design
- Delivered ready to use
- Tissues and cleaning fluid provided
The dispenser box is constructed of coated paperboard and has openings for wall mounting.
Lenses and cleaner are conveniently encased in dispenser. Tissues measure 12.7×20.3 cm (5×8"), packaged 600 count. Cleaning fluid is supplied in a 236.6 ml (8 oz.) container. Assembly not required.
Dispenses lens tissue and non-silicone cleaning fluid to protect, polish, antistat, and anti-fog lenses.