Lens Cleaning Solution, C-Clear
Fournisseur: C-Clear
Certificats
A propos de cet article
C-Clear lens cleaning solution is specially designed for the industrial setting.
- Contains no silicone
- Effectively cleans glass, plastic and polycarbonate lenses
- Used on sideshields, face shields, goggles, respirators and CRT screens
This environmentally safe fluid cuts through grease and oil while leaving an anti-fog/anti-static coating on the lenses.
Customers Who Bought This Also Bought