Thin Film Aniline Point Apparatus, Koehler
Fournisseur: Koehler
Certificats
A propos de cet article
Used to characterize pure hydrocarbons and to indicate the aromatic content of hydrocarbon mixtures according to Method B
Apparatus consists of a thin film tube, 400mL borosilicate glass beaker, cover assembly with bath stirrer, sample pump rotor and cooling coil, 6V lamp with line cord, and drive motor. A positive drive pulley system rotates sample and bath stirrers. Two thermometer ferrules and mounting hardware are included. Available Powertrol heaters provide variable stepless control and a reference dial for repeatable control of heating rate. They also provide a low voltage receptacle for the 6V lamp.
Unit stirs aniline sample mixture in a borosilicate glass thin film tube suspended in a heating bath.
Certifications: Conforms to specifications: ASTM D611, IP 2, ISO 2977, DIN 51775, FTM 791-3601.
Spécifications
- Catalog No:
- 37000-012
- 37000-010
- Dimensions:
- 37L x 22W x 53H cm (141/2 x 81/2 x 203/4")
- 37L x 22W x 53H cm (141/2 x 81/2 x 203/4")
- Shipping weight:
- 19.1 kg (42 lbs.)
- 19.1 kg (42 lbs.)