U-Tube Aniline Point Apparatus, Koehler
Fournisseur: Koehler
Certificats
A propos de cet article
Used to characterize pure hydrocarbons and to indicate the aromatic content of hydrocarbon mixtures according to IP2-56 Method D
The apparatus consists of a U-form aniline sample tube, 400mL borosilicate glass beaker, cover assembly with bath stirrer, sample stirrer, and cooling coil, 6V lamp with line cord, and drive motor. Unit includes thermometer ferrules and mounting hardware. Available Powertrol heaters provide variable stepless control of heating rate and a low voltage receptacle for the 6V lamp.
Suitable for samples having 6.5 or lighter ASTM D1500 color.
Certifications: Conforms to specifications: ASTM D611, IP 2, ISO 2977, DIN 51775, FTM 791-3601.
Spécifications
- Catalog No:
- 37000-008
- 37000-006
- Dimensions:
- 37L x 22W x 53H cm (141/2 x 81/2 x 203/4")
- 37L x 22W x 53H cm (141/2 x 81/2 x 203/4")
- Shipping weight:
- 19.1 kg (42 lbs.)
- 19.1 kg (42 lbs.)